Assay Details
Target Gene Details
Entrez Gene ID: | 67426 |
Gene Name: | aarF domain containing kinase 3 |
Gene Aliases: |
4632432J16Rik, AI462003, Cabc1, mKIAA0451 |
Location: |
Chr.1:180165238-180196020 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adck3 | NM_001163290.1 | 3 | 310 | NP_001156762.1 |
| NM_023341.3 | 3 | 408 | NP_075830.2 | |
| AJ278735.1 | 3 | 290 | CAC00538.1 | |
| AK004791.1 | 3 | 350 | BAB23567.2 | |
| AK014605.2 | 3 | 350 | BAB29459.2 | |
| AK179829.1 | 2 | 104 | ||
| AK179958.1 | 2 | 104 | ||
| BC030937.1 | 3 | 356 | AAH30937.1 | |
| BY261815.1 | ||||
| BY275962.1 |

Genomic Map