Assay Details
Target Gene Details
Entrez Gene ID: | 12912 |
Gene Name: | cAMP responsive element binding protein 1 |
Gene Aliases: |
2310001E10Rik, 3526402H21Rik, AV083133, Creb, Creb-1 |
Location: |
Chr.1:64532804-64604548 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Creb1 | NM_001037726.1 | 7 | 7033 | NP_001032815.1 |
NM_009952.2 | 9 | 7197 | NP_034082.1 | |
NM_133828.2 | 8 | 7155 | NP_598589.2 |
