Assay Details
Target Gene Details
Entrez Gene ID: | 26381 |
Gene Name: | estrogen-related receptor gamma |
Gene Aliases: |
ERR3, Errg, NR3B3, mKIAA0832 |
Location: |
Chr.1:187608792-188214884 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esrrg | NM_001243792.1 | 8 | 2023 | NP_001230721.1 |
NM_011935.3 | 7 | 1961 | NP_036065.1 | |
AB093266.1 | 8 | 2042 | BAC41450.1 | |
AK052731.1 | 8 | 2002 | BAC35120.1 | |
AK140676.1 | 5 | 1310 | BAE24441.1 | |
BC082324.1 | 7 | 1929 | AAH82324.1 |
