Assay Details
Target Gene Details
Entrez Gene ID: | 14048 |
Gene Name: | eyes absent 1 homolog (Drosophila) |
Gene Aliases: |
bor |
Location: |
Chr.1:14168953-14310265 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eya1 | NM_001252192.1 | 1 | 458 | NP_001239121.1 |
NM_010164.2 | 1 | 458 | NP_034294.2 | |
AK042412.1 | 1 | 381 | BAC31251.1 | |
AK045320.1 | 1 | 311 | ||
AK153788.1 | 1 | 463 | ||
BC060260.1 | 1 | 458 | AAH60260.1 | |
BC066860.1 | 1 | 115 |
