Assay Details
Target Gene Details
Entrez Gene ID: | 14768 |
Gene Name: | LanC (bacterial lantibiotic synthetase component C)-like 1 |
Gene Aliases: |
AW124738, Gpr69, Gpr69a, p40 |
Location: |
Chr.1:67000517-67038872 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lancl1 | NM_001190984.1 | 10 | 3237 | NP_001177913.1 |
| NM_001190985.1 | 9 | 3455 | NP_001177914.1 | |
| NM_021295.3 | 10 | 3271 | NP_067270.1 | |
| AJ294535.1 | 10 | 3264 | CAC69241.1 | |
| AK158930.1 | 1 | 464 |

Genomic Map