Assay Details
Target Gene Details
| Entrez Gene ID: | 69169 | 
| Gene Name: | Fc fragment of IgM receptor | 
| Gene Aliases: | 1810037B05Rik, Faim3, FcmuR, Toso | 
| Location: | Chr.1:130865777-130880790 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 8 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Fcmr | NM_026976.2 | 8 | 1378 | NP_081252.1 | 
| AK007714.1 | 8 | 1379 | BAB25207.1 | |
| AK155943.1 | 8 | 1379 | BAE33514.1 | |
| AY227385.1 | ||||
| AY227386.1 | ||||
| AY227387.1 | ||||
| AY227388.1 | ||||
| AY227389.1 | ||||
| AY227390.1 | ||||
| AY227391.1 | ||||
| AY227392.1 | ||||
| AY227393.1 | ||||
| BC116674.1 | 8 | 1483 | ||
| BC118505.1 | 8 | 1483 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top