Assay Details
Target Gene Details
Entrez Gene ID: | 211383 |
Gene Name: | APC membrane recruitment 3 |
Gene Aliases: |
9430069J07Rik, Fam123c |
Location: |
Chr.1:34578935-34590948 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amer3 | NM_213727.2 | 2 | 2214 | NP_998892.1 |
| AK034979.1 | 1 | 2086 | BAC28901.1 | |
| BC070435.1 | 2 | 2180 | AAH70435.1 |

Genomic Map