Assay Details
Target Gene Details
| Entrez Gene ID: | 18071 | 
| Gene Name: | nescient helix loop helix 1 | 
| Gene Aliases: | Hen1, Nscl, Tal2, bHLHa35 | 
| Location: | Chr.1:172052292-172057596 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Nhlh1 | NM_010916.2 | 2 | 1771 | NP_035046.1 | 
| AK149315.1 | BAE28808.1 | |||
| BC051018.1 | 2 | 1746 | AAH51018.1 | |
| M82874.1 | 2 | 1739 | AAA39840.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top