Assay Details
Target Gene Details
Entrez Gene ID: | 12370 |
Gene Name: | caspase 8 |
Gene Aliases: |
CASP-8, FLICE, MACH, Mch5 |
Location: |
Chr.1:58795229-58847503 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casp8 | NM_009812.2 | 1 | 180 | NP_033942.1 |
| AF067834.1 | 1 | 60 | AAC40131.1 | |
| AK146758.1 | 1 | 180 | BAE27412.1 | |
| AK168812.1 | 1 | 180 | BAE40639.1 | |
| BC006737.1 | 1 | 122 | AAH06737.1 | |
| BC049955.1 | AAH49955.1 |
More Information
Set Membership: |
|

Genomic Map