Assay Details
Target Gene Details
Entrez Gene ID: | 63913 |
Gene Name: | family with sequence similarity 129, member A |
Gene Aliases: |
AI256368, AU019833, Niban |
Location: |
Chr.1:151571373-151721939 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam129a | NM_022018.3 | 14 | 2872 | NP_071301.2 |
| AB049355.1 | 14 | 2712 | BAB17052.1 | |
| AK029847.1 | 10 | 2184 | BAC26641.1 | |
| AK136597.1 | 14 | 2782 | BAE23066.1 | |
| AK170193.1 | 15 | 2950 | BAE41626.1 | |
| AK170347.1 | 5 | 1467 | BAE41736.1 | |
| AK187603.1 | 1 | 170 | ||
| AK191335.1 | 1 | 170 | ||
| AK199589.1 | 1 | 167 | ||
| AK209783.1 | 1 | 170 | ||
| BC058234.1 | 12 | 2472 | ||
| BC137843.1 | 10 | 2249 |

Genomic Map