Assay Details
Target Gene Details
Entrez Gene ID: | 226438 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
9830123M21Rik |
Location: |
Chr.1:135953205-136040842 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igfn1 | NM_177642.3 | 22 | 7375 | NP_808310.2 |
AK036509.1 | 3 | 256 | BAC29455.1 | |
EU429481.1 | 8 | 4247 | ||
EU429482.1 | 7 | 1114 | ||
EU429483.2 | 17 | 2528 | ||
GU290042.1 | 22 | 7375 |
