Assay Details
Target Gene Details
Entrez Gene ID: | 226438 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
9830123M21Rik |
Location: |
Chr.1:135953205-136040842 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igfn1 | NM_177642.3 | 26 | 8430 | NP_808310.2 |
AK036509.1 | 7 | 1205 | BAC29455.1 | |
BC107027.1 | 4 | 754 | AAI07028.1 | |
BC107028.2 | 4 | 754 | AAI07029.1 | |
EU429481.1 | 12 | 5302 | ||
EU429482.1 | 11 | 2169 | ||
EU429483.2 | 21 | 3583 | ||
GU290042.1 | 26 | 8430 |
