Assay Details
Target Gene Details
Entrez Gene ID: | 140559 |
Gene Name: | immunoglobulin superfamily, member 8 |
Gene Aliases: |
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384 |
Location: |
Chr.1:172312407-172319841 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf8 | NM_080419.1 | 7 | 2065 | NP_536344.1 |
| AF411055.1 | 7 | 1936 | AAL02217.1 | |
| AF439263.1 | 7 | 2048 | AAN63626.1 | |
| AK140254.1 | 7 | 2077 | ||
| AK146104.1 | 7 | 2078 | BAE26904.1 | |
| AK146221.1 | 7 | 2084 | BAE26989.1 | |
| AK158522.1 | 7 | 2086 | BAE34544.1 | |
| AK168363.1 | 7 | 2084 | BAE40298.1 | |
| AK186312.1 | 2 | 106 | ||
| AK189184.1 | 3 | 248 | ||
| AK190325.1 | 2 | 211 | ||
| AK190456.1 | 2 | 106 | ||
| AK193134.1 | 2 | 111 | ||
| AK193634.1 | 2 | 106 | ||
| AK193780.1 | 2 | 106 | ||
| AK196050.1 | 2 | 106 | ||
| AK198924.1 | 2 | 106 | ||
| AK200236.1 | 2 | 107 | ||
| AK200799.1 | 2 | 106 | ||
| AK200990.1 | 2 | 106 | ||
| AK201824.1 | 2 | 106 | ||
| AK205206.1 | 2 | 106 | ||
| AK207017.1 | 2 | 106 | ||
| AK208609.1 | 1 | 99 | ||
| AK209167.1 | 2 | 106 | ||
| AK210016.1 | 2 | 106 | ||
| AK212259.1 | 1 | 106 | ||
| AK216251.1 | 2 | 106 | ||
| AK216541.1 | 2 | 218 | ||
| AK216980.1 | 2 | 106 | ||
| BC019587.1 | 5 | 1087 | AAH19587.1 | |
| BC026464.1 | 6 | 1871 | AAH26464.1 | |
| BC026633.1 | 3 | 617 | AAH26633.1 | |
| BC048387.1 | 7 | 2065 | AAH48387.1 |
More Information
Set Membership: |
|

Genomic Map