Assay Details
Target Gene Details
Entrez Gene ID: | 68421 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
0910001K20Rik, AV347960 |
Location: |
Chr.1:24678544-24766301 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmbrd1 | NM_026719.2 | 16 | 2939 | NP_080995.2 |
| AK033437.1 | 1 | 508 | ||
| AK152170.1 | 16 | 2750 | BAE31002.1 | |
| AK153237.1 | 16 | 2750 | BAE31829.1 | |
| AK158982.1 | 16 | 2775 | BAE34756.1 | |
| BC025579.1 | 16 | 2771 | AAH25579.1 |
More Information
Set Membership: |
|

Genomic Map