Assay Details
Target Gene Details
Entrez Gene ID: | 226304 |
Gene Name: | neuropeptides B/W receptor 1 |
Gene Aliases: |
Gpr7, Nbpwr1 |
Location: |
Chr.1:5913707-5917398 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npbwr1 | NM_010342.1 | 1 | 1850 | NP_034472.1 |
| AK138357.1 | 1 | 1851 |
More Information
Set Membership: |
|

Genomic Map