Assay Details
Target Gene Details
Entrez Gene ID: | 319455 |
Gene Name: | phospholipase D family, member 5 |
Gene Aliases: |
B230365F16Rik |
Location: |
Chr.1:175962301-176280017 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pld5 | NM_176916.4 | 1 | 523 | NP_795890.2 |
| AK043287.1 | 1 | 486 | BAC31515.1 | |
| AK046294.1 | 1 | 486 | ||
| AK136851.1 | 1 | 524 | BAE23146.1 | |
| AK136888.1 | 1 | 524 | BAE23156.1 | |
| AK144116.1 | 1 | 512 | BAE25709.1 |
More Information
Set Membership: |
|

Genomic Map