Assay Details
Target Gene Details
Entrez Gene ID: | 69171 |
Gene Name: | cyclin Pas1/PHO80 domain containing 1 |
Gene Aliases: |
1810031K17Rik, AU043960 |
Location: |
Chr.1:75135215-75142733 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnppd1 | NM_026977.2 | 9 | 1309 | NP_081253.1 |
AK007667.1 | 1 | 196 | BAB25174.1 | |
AK029090.1 | 9 | 1309 | BAC26289.1 | |
AK080437.1 | 3 | 1527 | ||
AK145962.1 | 10 | 1556 | BAE26787.1 | |
AK169779.1 | 9 | 1581 | BAE41361.1 | |
AK181449.1 | 1 | 301 | ||
AK209810.1 | 1 | 301 | ||
AK210191.1 | 1 | 254 | ||
BC013714.1 | 8 | 1082 | ||
BC028815.1 | 9 | 1294 | AAH28815.1 |
