Assay Details
Target Gene Details
Entrez Gene ID: | 72949 |
Gene Name: | cyclin T2 |
Gene Aliases: |
2900041I18Rik, C81304, CycT2, CycT2a, CycT2b |
Location: |
Chr.1:127774164-127804837 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccnt2 | NM_028399.1 | 9 | 2173 | NP_082675.1 |
AK047125.1 | 5 | 3073 | ||
AK049863.1 | 10 | 2020 | ||
AK152399.1 | 9 | 2121 | BAE31187.1 | |
AK154439.1 | 9 | 2131 | BAE32588.1 | |
BC054122.1 | 9 | 2173 | AAH54122.1 | |
JX965958.1 | 9 | 2109 |
