Assay Details
Target Gene Details
Entrez Gene ID: | 226971 |
Gene Name: | pleckstrin homology domain containing, family B (evectins) member 2 |
Gene Aliases: |
2310009M15Rik, AU018239, Evt2, Phdc, evt-2 |
Location: |
Chr.1:34849959-34879585 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhb2 | NM_145516.2 | 8 | 1981 | NP_663491.1 |
| AF189817.1 | 8 | 1941 | AAF01332.1 | |
| AK039771.1 | 8 | 1949 | BAC30448.1 | |
| AK166091.1 | 8 | 1949 | BAE38565.1 | |
| AK181418.1 | 1 | 162 | ||
| AK190116.1 | 1 | 156 | ||
| AK213647.1 | 1 | 156 | ||
| BC057994.1 | 10 | 1342 | AAH57994.1 |

Genomic Map