Assay Details
Target Gene Details
Entrez Gene ID: | 18986 |
Gene Name: | POU domain, class 2, transcription factor 1 |
Gene Aliases: |
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1 |
Location: |
Chr.1:165865150-166002672 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pou2f1 | NM_011137.3 | 17 | 4967 | NP_035267.2 |
NM_198932.3 | 16 | 4895 | NP_945150.2 | |
NM_198933.3 | 16 | 4967 | NP_945151.2 | |
NM_198934.3 | 17 | 5039 | NP_945152.2 |
