Assay Details
Target Gene Details
Entrez Gene ID: | 12503 |
Gene Name: | CD247 antigen |
Gene Aliases: |
4930549J05Rik, A430104F18Rik, AW552088, Cd3, Cd3-eta, Cd3-zeta, Cd3h, Cd3z, Cd3zeta, T3z, Tcrk, Tcrz |
Location: |
Chr.1:165780357-165871153 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd247 | NM_001113392.2 | NP_001106863.1 | ||
| NM_001113393.2 | NP_001106864.2 | |||
| NM_031162.4 | 8 | 1227 | NP_112439.1 | |
| NR_103716.1 | ||||
| AK138774.1 | BAE23774.1 | |||
| AK139276.1 | 1 | 2693 | ||
| BF020526.1 | 1 | 140 | ||
| L03353.1 | AAA37401.1 | |||
| M33158.1 | 8 | 1217 | AAA37398.1 | |
| S51932.1 | AAB24559.1 | |||
| U56258.1 | AAD11545.1 | |||
| X84237.1 | CAA59015.1 |
Target Gene Details
Entrez Gene ID: | 18986 |
Gene Name: | POU domain, class 2, transcription factor 1 |
Gene Aliases: |
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1 |
Location: |
Chr.1:165865150-166002672 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pou2f1 | NM_011137.3 | 17 | 10828 | NP_035267.2 |
| NM_198932.3 | 16 | 10756 | NP_945150.2 | |
| NM_198933.3 | 16 | 10828 | NP_945151.2 | |
| NM_198934.3 | 17 | 10900 | NP_945152.2 |
More Information
Set Membership: |
|

Genomic Map