Assay Details
Target Gene Details
Entrez Gene ID: | 16980 |
Gene Name: | leucine rich repeat protein 2, neuronal |
Gene Aliases: |
5730406J09Rik, NLRR-2 |
Location: |
Chr.1:132880355-132940005 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrn2 | NM_010732.4 | 1 | 154 | NP_034862.1 |
| AK036271.1 | 1 | 155 | ||
| AK039216.1 | 1 | 155 |
More Information
Set Membership: |
|

Genomic Map