Assay Details
Target Gene Details
Entrez Gene ID: | 18187 |
Gene Name: | neuropilin 2 |
Gene Aliases: |
1110048P06Rik, Np-2, Np2, Npn-2, Npn2 |
Location: |
Chr.1:62703317-62818692 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrp2 | NM_001077403.1 | 17 | 4496 | NP_001070871.1 |
| NM_001077404.1 | 17 | 4481 | NP_001070872.1 | |
| NM_001077405.1 | 16 | 4430 | NP_001070873.1 | |
| NM_010939.2 | 16 | 4445 | NP_035069.2 | |
| BC029876.1 | 8 | 2198 | AAH29876.1 |

Genomic Map