Assay Details
Target Gene Details
Entrez Gene ID: | 71281 |
Gene Name: | apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 4 (putative) |
Gene Aliases: |
4933431M11Rik |
Location: |
Chr.1:152750551-152757544 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apobec4 | NM_001081197.1 | 2 | 656 | NP_001074666.1 |
| AK017014.1 | 2 | 631 | ||
| BC100465.1 | 2 | 656 | AAI00466.1 |
Target Gene Details
Entrez Gene ID: | 19731 |
Gene Name: | ral guanine nucleotide dissociation stimulator,-like 1 |
Gene Aliases: |
Rgl, mKIAA0959 |
Location: |
Chr.1:152516768-152766351 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgl1 |

Genomic Map