Assay Details
Target Gene Details
Entrez Gene ID: | 14125 |
Gene Name: | Fc receptor, IgE, high affinity I, alpha polypeptide |
Gene Aliases: |
FcERI, Fce1a, Fcr-5, fcepsilonri |
Location: |
Chr.1:173221269-173227232 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcer1a | NM_010184.2 | 3 | 297 | NP_034314.2 |
| BC125455.1 | 3 | 262 | ||
| BC125457.1 | 3 | 261 | ||
| CJ142154.1 | 3 | 218 | ||
| CJ142191.1 | 3 | 297 | ||
| J05018.1 | 3 | 282 | AAA37600.1 |
More Information
Set Membership: |
|

Genomic Map