Assay Details
Target Gene Details
Entrez Gene ID: | 76709 |
Gene Name: | actin related protein 2/3 complex, subunit 2 |
Gene Aliases: |
2210023N03Rik, 34kDa, p34-Arc |
Location: |
Chr.1:74236504-74268213 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arpc2 | NM_029711.1 | 1 | 231 | NP_083987.1 |
AK151513.1 | 1 | 205 | BAE30463.1 | |
AK152356.1 | BAE31148.1 | |||
AK170061.1 | ||||
BC115750.1 | AAI15751.1 | |||
BF016986.1 | 1 | 231 |
