Assay Details
Target Gene Details
Entrez Gene ID: | 76709 |
Gene Name: | actin related protein 2/3 complex, subunit 2 |
Gene Aliases: |
2210023N03Rik, 34kDa, p34-Arc |
Location: |
Chr.1:74236504-74268213 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arpc2 | NM_029711.1 | 10 | 1297 | NP_083987.1 |
AK083263.1 | 10 | 1107 | ||
AK151513.1 | 10 | 1271 | BAE30463.1 | |
AK152356.1 | 11 | 1122 | BAE31148.1 | |
AK170061.1 | 11 | 1096 | ||
AK213157.1 | 1 | 73 | ||
BC099504.1 | 9 | 949 | AAH99504.1 | |
BY440113.1 | 1 | 102 |
