Assay Details
Target Gene Details
Entrez Gene ID: | 67771 |
Gene Name: | actin related protein 2/3 complex, subunit 5 |
Gene Aliases: |
5830443F10Rik, p16-Arc |
Location: |
Chr.1:152766542-152775580 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpc5 | NM_026369.2 | 1 | 105 | NP_080645.2 |
| AK008097.1 | BAB25457.1 | |||
| AK011348.1 | BAB27558.1 | |||
| AK011911.1 | BAB27911.1 | |||
| AK017669.1 | BAB30864.1 | |||
| AK017985.1 | ||||
| AK019149.1 | BAB31570.1 | |||
| AK087427.1 | BAC39870.1 | |||
| AK149756.1 | BAE29064.1 | |||
| AK150443.1 | BAE29565.1 | |||
| AK150703.1 | BAE29782.1 | |||
| AK151490.1 | BAE30443.1 | |||
| AK151654.1 | BAE30583.1 | |||
| AK151656.1 | BAE30585.1 | |||
| AK152281.1 | BAE31095.1 | |||
| AK153262.1 | BAE31852.1 | |||
| AK159407.1 | BAE35058.1 | |||
| AK159438.1 | BAE35084.1 | |||
| AK159738.1 | BAE35332.1 | |||
| AK166284.1 | BAE38681.1 | |||
| AK166335.1 | BAE38714.1 | |||
| AK166725.1 | BAE38972.1 | |||
| AK166919.1 | BAE39116.1 | |||
| AK167916.1 | BAE39923.1 | |||
| AK168264.1 | BAE40212.1 | |||
| BC060143.1 | AAH60143.1 | |||
| CF170085.1 | 1 | 105 |
More Information
Set Membership: |
|

Genomic Map