Assay Details
Target Gene Details
Entrez Gene ID: | 12570 |
Gene Name: | cyclin-dependent kinase 5, regulatory subunit 2 (p39) |
Gene Aliases: |
B230310J22Rik, p39 |
Location: |
Chr.1:74855029-74857732 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5r2 | NM_009872.3 | 1 | 1796 | NP_034002.2 |
| AK080815.1 |
More Information
Set Membership: |
|

Genomic Map