Assay Details
Target Gene Details
Entrez Gene ID: | 319901 |
Gene Name: | dermatan sulfate epimerase-like |
Gene Aliases: |
9330132E09Rik, DS-epi2 |
Location: |
Chr.1:111858701-111864918 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dsel | NM_001081316.1 | 2 | 3305 | NP_001074785.1 |
AK032110.1 | 1 | 1195 | ||
AK033983.1 | 2 | 3306 | BAC28538.1 | |
AK164854.1 | ||||
BC120567.1 | 1 | 1667 | ||
BC137796.1 | 1 | 1667 |
