Assay Details
Target Gene Details
Entrez Gene ID: | 55990 |
Gene Name: | flavin containing monooxygenase 2 |
Gene Aliases: |
2310008D08Rik, 2310042I22Rik, AW107733 |
Location: |
Chr.1:162874321-162898712 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fmo2 | NM_018881.3 | 7 | 1112 | NP_061369.2 |
| AF184981.1 | 6 | 1177 | AAD56413.1 | |
| AK129026.1 | 3 | 645 | ||
| BC031415.1 | 7 | 1112 | AAH31415.1 |
More Information
Set Membership: |
|

Genomic Map