Assay Details
Target Gene Details
Entrez Gene ID: | 100125339 |
Gene Name: | RIKEN cDNA A230020J21 gene |
Gene Aliases: |
ENSMUSG00000066596 |
Location: |
Chr.1:191025351-191029781 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A230020J21Rik | NR_027298.1 | 1 | 157 | |
| AK038499.1 | 1 | 158 | BAC30019.1 |
Target Gene Details
Entrez Gene ID: | 226844 |
Gene Name: | major facilitator superfamily domain containing 7B |
Gene Aliases: |
9630055N22Rik, FLVCR, Flvcr1 |
Location: |
Chr.1:191005832-191026215 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd7b | NM_001081259.1 | 1 | 684 | NP_001074728.1 |
| AK036315.1 | 1 | 462 | ||
| AK165872.1 | 1 | 683 | ||
| BC157992.1 | 1 | 652 |
More Information
Set Membership: |
|

Genomic Map