Assay Details
Target Gene Details
Entrez Gene ID: | 19070 |
Gene Name: | MOB family member 4, phocein |
Gene Aliases: |
2610109B12Rik, Mobkl3, Phocn, Prei3 |
Location: |
Chr.1:55130936-55154899 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mob4 | NM_025283.3 | 8 | 1344 | NP_079559.2 |
| AK147857.1 | 8 | 1303 | BAE28182.1 | |
| AK162327.1 | 8 | 1331 | BAE36855.1 | |
| AK185109.1 | 1 | 368 | ||
| BC058168.1 | 8 | 1297 | AAH58168.1 |
More Information
Set Membership: |
|

Genomic Map