Assay Details
Target Gene Details
Entrez Gene ID: | 20725 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 8 |
Gene Aliases: |
CAP-2, CAP2, NK10, Spi8, ovalbumin |
Location: |
Chr.1:107589925-107610472 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb8 | NM_011459.4 | 7 | 2047 | NP_035589.1 |
| AK032659.1 | 7 | 2046 | ||
| AK036811.1 | 7 | 1983 | BAC29588.1 | |
| AK148413.1 | 7 | 2045 | BAE28539.1 |
More Information
Set Membership: |
|

Genomic Map