Assay Details
Target Gene Details
Entrez Gene ID: | 68481 |
Gene Name: | myelin protein zero-like 1 |
Gene Aliases: |
1110007A10Rik, PZR |
Location: |
Chr.1:165592181-165634558 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpzl1 | NM_001001880.2 | 5 | 1260 | NP_001001880.1 |
| NM_001083897.1 | 6 | 1366 | NP_001077366.1 | |
| AK003513.1 | 1 | 318 | ||
| AK090278.1 | 6 | 1352 | ||
| AK169385.1 | 6 | 1356 | BAE41132.1 | |
| AK188720.1 | 1 | 65 | ||
| AK198363.1 | 1 | 65 | ||
| AK200888.1 | 1 | 65 | ||
| AK206772.1 | 1 | 65 | ||
| AK210071.1 | 1 | 65 | ||
| AK214871.1 | 1 | 65 | ||
| AK215062.1 | 1 | 65 | ||
| AK218959.1 | 1 | 65 | ||
| AY764247.1 | 6 | 1372 | AAX11678.1 | |
| BC072563.1 | 5 | 1255 | AAH72563.1 |
More Information
Set Membership: |
|

Genomic Map