Assay Details
Target Gene Details
Entrez Gene ID: | 210933 |
Gene Name: | adhesion G protein-coupled receptor B3 |
Gene Aliases: |
A830096D10Rik, Bai3 |
Location: |
Chr.1:25067476-25830206 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgrb3 | NM_175642.4 | 31 | 4946 | NP_783573.4 |
| AK044248.1 | 16 | 2380 | BAC31839.1 | |
| AK081421.1 | 10 | 2288 | BAC38215.1 | |
| BC032251.1 | 24 | 3163 | AAH32251.1 | |
| BC079670.1 | 29 | 3965 | AAH79670.1 | |
| BC099951.1 | 31 | 4944 | AAH99951.1 |
More Information
Set Membership: |
|

Genomic Map