Assay Details
Target Gene Details
Entrez Gene ID: | 69601 |
Gene Name: | disabled 2 interacting protein |
Gene Aliases: |
2310011D08Rik, AI480459, Aip1, mKIAA1743 |
Location: |
Chr.2:35558458-35730994 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dab2ip | NM_001001602.2 | 10 | 2727 | NP_001001602.2 |
| NM_001114124.2 | 12 | 2851 | NP_001107596.1 | |
| NM_001114125.1 | 12 | 2703 | NP_001107597.1 | |
| NM_001290637.1 | 12 | 2758 | NP_001277566.1 | |
| NM_001290639.1 | 10 | 2386 | NP_001277568.1 | |
| NM_001290640.1 | 10 | 2691 | NP_001277569.1 | |
| NM_001290641.1 | 12 | 2984 | NP_001277570.1 | |
| AK122548.1 | 7 | 1691 | BAC65830.1 | |
| AK147464.1 | 12 | 2783 | BAE27930.1 | |
| AK147593.1 | 12 | 2985 | BAE28013.1 | |
| AK155296.1 | 12 | 2759 | BAE33173.1 | |
| AY178784.1 | 2 | 619 | AAP31233.1 | |
| AY305656.1 | 10 | 2444 | AAQ77379.1 | |
| AY305657.1 | 10 | 2727 | AAQ77380.1 | |
| AY305658.1 | 10 | 2386 | AAQ77381.1 | |
| BC094373.1 | 7 | 1482 | AAH94373.1 | |
| BC118530.1 | 10 | 2352 | ||
| DQ473307.1 | 12 | 2588 |
More Information
Set Membership: |
|

Genomic Map