Assay Details
Target Gene Details
Entrez Gene ID: | 54338 |
Gene Name: | solute carrier family 23 (nucleobase transporters), member 2 |
Gene Aliases: |
AI844736, NBTL1, SVCT2, Slc23a1, YSPL2, YSPL3, mKIAA0238 |
Location: |
Chr.2:132052496-132145108 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc23a2 | NM_018824.2 | 16 | 5637 | NP_061294.2 |
| AK122227.1 | 12 | 5409 | BAC65509.1 | |
| AK163600.1 | 1 | 2654 | ||
| AY004874.1 | 16 | 5591 | AAG02252.1 | |
| BC050823.1 | 15 | 5401 | AAH50823.1 |
More Information
Set Membership: |
|

Genomic Map