Assay Details
Target Gene Details
Entrez Gene ID: | 11350 |
Gene Name: | c-abl oncogene 1, non-receptor tyrosine kinase |
Gene Aliases: |
AI325092, Abl, E430008G22Rik, c-Abl |
Location: |
Chr.2:31688252-31807093 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abl1 | NM_001112703.2 | 1 | 482 | NP_001106174.1 |
NM_001283046.1 | 1 | 482 | NP_001269975.1 | |
NM_001283047.1 | 1 | 482 | NP_001269976.1 | |
AK083248.1 | 1 | 644 | ||
AK088252.1 | 1 | 468 | BAE43394.1 | |
AK090247.1 | 1 | 483 | ||
AK166028.1 | 1 | 469 | ||
AK166210.1 | 1 | 610 | BAE38629.1 |
