Assay Details
Target Gene Details
Entrez Gene ID: | 12561 |
Gene Name: | cadherin 4 |
Gene Aliases: |
AW120700, R-Cadh, R-cadherin, Rcad |
Location: |
Chr.2:179442478-179899377 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh4 | NM_009867.2 | 16 | 3897 | NP_033997.1 |
| AK038529.1 | 1 | 620 | ||
| AK049087.1 | 1 | 556 |
More Information
Set Membership: |
|

Genomic Map