Assay Details
Target Gene Details
Entrez Gene ID: | 52840 |
Gene Name: | dysbindin (dystrobrevin binding protein 1) domain containing 2 |
Gene Aliases: |
1110017A21Rik, 2900022J10Rik, AU041050, AW048677, D2Bwg0891e, Nkip, R74724 |
Location: |
Chr.2:164472235-164493323 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbndd2 | NM_001048227.1 | NP_001041692.1 | ||
| NM_001048228.1 | NP_001041693.1 | |||
| NM_001048229.1 | 1 | 142 | NP_001041694.1 | |
| NM_026797.2 | NP_081073.1 | |||
| AK003735.1 | BAB22969.1 | |||
| AK013571.1 | BAB28911.1 | |||
| AY187289.1 | AAO64484.1 | |||
| BC019947.1 | AAH19947.1 | |||
| BC030298.1 | AAH30298.1 | |||
| BG080686.2 | ||||
| CJ156083.1 | 1 | 144 | ||
| CX241653.1 |

Genomic Map