Assay Details
Target Gene Details
Entrez Gene ID: | 52840 |
Gene Name: | dysbindin (dystrobrevin binding protein 1) domain containing 2 |
Gene Aliases: |
1110017A21Rik, 2900022J10Rik, AU041050, AW048677, D2Bwg0891e, Nkip, R74724 |
Location: |
Chr.2:164472235-164493323 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbndd2 | NM_001048227.1 | 4 | 3036 | NP_001041692.1 |
| NM_001048228.1 | 4 | 3060 | NP_001041693.1 | |
| NM_001048229.1 | 3 | 3063 | NP_001041694.1 | |
| NM_026797.2 | 4 | 3165 | NP_081073.1 | |
| AK045608.1 | 1 | 1132 |

Genomic Map