Assay Details
Target Gene Details
Entrez Gene ID: | 14049 |
Gene Name: | eyes absent 2 homolog (Drosophila) |
Gene Aliases: |
Eab1 |
Location: |
Chr.2:165595028-165771727 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eya2 | NM_001271962.1 | 16 | 1883 | NP_001258891.1 |
NM_001271963.1 | 16 | 2059 | NP_001258892.1 | |
NM_010165.3 | 16 | 2060 | NP_034295.1 | |
AK137718.1 | 16 | 2055 | BAE23473.1 | |
AK141765.1 | 16 | 2059 | BAE24826.1 | |
BC003755.1 | 16 | 2060 | AAH03755.1 | |
U61111.1 | 15 | 1801 | AAB48018.1 | |
U71208.1 | 16 | 1885 | AAB51121.1 | |
U81603.1 | 15 | 1800 | AAB42067.1 |
