Assay Details
Target Gene Details
Entrez Gene ID: | 241638 |
Gene Name: | leucine zipper, putative tumor suppressor family member 3 |
Gene Aliases: |
Prosapip1 |
Location: |
Chr.2:130632765-130642844 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lzts3 | NM_001291027.1 | 4 | 1053 | NP_001277956.1 |
| NM_001291028.1 | 4 | 711 | NP_001277957.1 | |
| NM_197945.4 | 4 | 1053 | NP_922936.3 | |
| AK148251.1 | 4 | 1012 | ||
| AK161711.1 | 4 | 1007 | BAE36545.1 | |
| BC058280.1 | 3 | 635 | AAH58280.1 | |
| DN175861.1 |

Genomic Map