Assay Details
Target Gene Details
Entrez Gene ID: | 170441 |
Gene Name: | solute carrier family 2 (facilitated glucose transporter), member 10 |
Gene Aliases: |
AA450473, Glut10 |
Location: |
Chr.2:165503897-165519917 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc2a10 | NM_130451.3 | 2 | 572 | NP_569718.1 |
| AK052156.1 | 2 | 539 | BAC34861.1 | |
| AY029579.1 | 2 | 495 | AAK38739.1 | |
| BC137917.1 | 2 | 490 | ||
| BC137920.1 | 2 | 490 |

Genomic Map