Assay Details
Target Gene Details
Entrez Gene ID: | 12048 |
Gene Name: | BCL2-like 1 |
Gene Aliases: |
Bcl(X)L, Bcl-XL, Bcl2l, BclX, bcl-x, bcl2-L-1 |
Location: |
Chr.2:152758732-152832581 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l1 | NM_001289716.1 | 2 | 781 | NP_001276645.1 |
| NM_001289717.1 | 2 | 626 | NP_001276646.1 | |
| NM_001289739.1 | 2 | 781 | NP_001276668.1 | |
| NM_009743.5 | 2 | 575 | NP_033873.3 | |
| AK146461.1 | 2 | 731 | BAE27189.1 | |
| AK150028.1 | 2 | 517 | ||
| AK172250.1 | 2 | 542 | BAE42906.1 | |
| BC089016.1 | 2 | 529 | AAH89016.1 | |
| BC089017.1 | 1 | 366 | AAH89017.1 | |
| L35049.1 | 2 | 580 | AAA51039.1 | |
| U10100.1 | AAA82172.1 | |||
| U10101.1 | 1 | 337 | AAA82173.1 | |
| U10102.1 | 2 | 574 | AAA82174.1 | |
| U51277.1 | 2 | 714 | AAC53458.1 | |
| U51278.1 | 1 | 439 | AAC53459.1 | |
| U51279.1 | 2 | 572 | AAC53460.1 | |
| X83574.1 | 1 | 337 | CAA58557.1 |

Genomic Map