Assay Details
Target Gene Details
Entrez Gene ID: | 12048 |
Gene Name: | BCL2-like 1 |
Gene Aliases: |
Bcl(X)L, Bcl-XL, Bcl2l, BclX, bcl-x, bcl2-L-1 |
Location: |
Chr.2:152758732-152832581 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl2l1 | NM_001289716.1 | 3 | 1551 | NP_001276645.1 |
| NM_001289717.1 | 3 | 1396 | NP_001276646.1 | |
| NM_001289739.1 | NP_001276668.1 | |||
| NM_009743.5 | 3 | 1345 | NP_033873.3 | |
| AK146461.1 | 3 | 1501 | BAE27189.1 | |
| AK150028.1 | 3 | 1288 | ||
| AK172250.1 | 3 | 1312 | BAE42906.1 | |
| AK178356.1 | ||||
| AK179733.1 | ||||
| AK206542.1 | ||||
| AK211933.1 | ||||
| AK211979.1 | ||||
| BC089016.1 | 3 | 1299 | AAH89016.1 | |
| BC089017.1 | 2 | 1136 | AAH89017.1 | |
| U51277.1 | AAC53458.1 | |||
| U51278.1 | 2 | 1209 | AAC53459.1 |

Genomic Map