Assay Details
Target Gene Details
Entrez Gene ID: | 319513 |
Gene Name: | PC-esterase domain containing 1A |
Gene Aliases: |
A930025D01Rik, AI480484, Fam113a |
Location: |
Chr.2:130417684-130424641 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pced1a | NM_001114541.1 | NP_001108013.1 | ||
NM_178762.4 | 1 | 585 | NP_848877.2 | |
AK027964.1 | 1 | 576 | ||
AK031469.1 | 1 | 999 | BAC27419.1 | |
AK032354.1 | 1 | 576 | BAC27830.1 | |
AK032762.1 | 1 | 560 | BAC28010.1 | |
AK041939.1 | 1 | 579 | ||
AK044590.1 | 1 | 570 | BAC31990.1 | |
AK045248.1 | 1 | 564 | BAC32279.1 | |
AK077745.1 | 1 | 576 | ||
AK081584.1 | 1 | 546 | BAC38264.1 | |
AK085613.1 | 1 | 579 | BAC39486.1 | |
AK161998.1 | 1 | 576 | ||
AK179623.1 | ||||
BB632143.1 | ||||
BC064802.1 | AAH64802.1 | |||
BY326318.1 |
