Assay Details
Target Gene Details
Entrez Gene ID: | 19124 |
Gene Name: | protein C receptor, endothelial |
Gene Aliases: |
AI325044, Ccca, Ccd41, Epcr |
Location: |
Chr.2:155751125-155755478 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Procr | NM_011171.2 | 2 | 420 | NP_035301.2 |
| AK136541.1 | 2 | 280 | ||
| AK143719.1 | 2 | 522 | BAE25514.1 | |
| AK167302.1 | 2 | 360 | BAE39407.1 | |
| BC028755.1 | 2 | 293 | AAH28755.1 | |
| CT010175.1 | 2 | 239 | CAJ18383.1 | |
| L39017.1 | 2 | 294 | AAC42049.1 | |
| X63748.2 | 2 | 326 | CAA45281.2 |
More Information
Set Membership: |
|

Genomic Map