Assay Details
Target Gene Details
Entrez Gene ID: | 68295 |
Gene Name: | AAR2 splicing factor homolog (S. cerevisiae) |
Gene Aliases: |
0610011L14Rik |
Location: |
Chr.2:156547558-156568972 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aar2 | NM_001164818.1 | 5 | 2888 | NP_001158290.1 |
| NM_026661.4 | 4 | 2745 | NP_080937.3 | |
| AK028890.1 | 4 | 2723 | BAC26177.1 |
More Information
Set Membership: |
|

Genomic Map